Order Entry
Spain
ContactUsLinkComponent
 
Sodium-L(+)-glutamate monohydrate ≥99.0% (by titrimetric analysis)
 :  TCI
 
undefined
Sodium-L(+)-glutamate monohydrate ≥99.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Specific rotation [a]20/D:
    +24.0 to +27.0 deg (C=10, 2mol/L HCl)
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together