(±)-Mandelic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
CAS Number:90-64-2
Formula:C₈H₈O₃
MDL Number:MFCD00064250
:152.15 g/mol
Merck Index:14,05717
Boiling Pt:322 °C (1013 hPa)
Melting Pt:117…120 °C
Density:1,361 g/cm³ (20 °C)
EINECS:202-007-6
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:119.0 to 122.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together