

Quinhydrone
Supplier: TCI
Certificates
About this item
CAS Number:106-34-3
:
Formula:C₆H₄(OH)₂·C₆H₄O₂
MDL Number:MFCD00010310
:218.21 g/mol
Merck Index:14,08058
Melting Pt:168…171 °C
Density:1,35 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-387-6
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Melting point:169.0 to 173.0 deg-C
- Solubility in EtOH:almost transparency
- p-Benzoquinone:48.0 to 52.0 %
- Hydroquinone:48.0 to 52.0 %
Specification Test Results
Frequently Bought Together