
4-Hydroxy-3-nitroanisole ≥98.0% (by GC, titration analysis)
: TCI
:1568-70-3
:
:C₇H₇NO₄
:MFCD00024247
:169.14 g/mol
:78…80 °C
:1,497 g/cm³ (20 °C)
:GHS07
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:78.0 to 82.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together