Methyl cinnamate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate
CAS Number:103-26-4
:
Formula:C₁₀H₁₀O₂
MDL Number:MFCD00008458
:162.19 g/mol
Merck Index:14,02299
Storage Temperature:Refrigerator
EINECS:203-093-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together