Benzoylcholine chloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAB0107-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
Melting Pt:210 °C
:243.73 g/mol
EINECS:221-000-9
:GHS07
CAS Number:2964-09-2
Formula:C₆H₅C(O)O(CH₂)₂N(CH₃)₃Cl
MDL Number:MFCD00011786