
3-Amino-2-cyclohexen-1-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5220-49-5
:
Formula:C₆H₉NO
MDL Number:MFCD00013783
:111.14 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:60 °C
EINECS:226-014-9
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 97.0 % (calcd.on anh.substance)
- Melting point:127.0 to 134.0 deg-C
- Water:max. 6.0 %
Specification Test Results
Frequently Bought Together