Order Entry
Spain
ContactUsLinkComponent
 
 
4-Biphenylylmethanol ≥99.0% (by GC)
  TCIAH0608-25G
 :  TCI
 :  
undefined
 
4-Biphenylylmethanol ≥99.0% (by GC)
  TCIAH0608-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    103.0 to 107.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :184.24 g/mol
 :184°C
 :104…105 °C
 :222-745-2
 :GHS08
 :3597-91-9
 :C₁₃H₁₂O
 :MFCD00004660