3,4-Dichlorobenzenesulphonyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:98-31-7
:
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00041255
:245.51 g/mol
Boiling Pt:158 - 160 °C (15 -rr)
Melting Pt:24.2…24.8 °C
Density:1,572 g/cm³ (20 °C)
EINECS:202-656-5
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Almost white wax
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together