
Silver trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
: TCI
:2923-28-6
:
:CAgF₃O₃S
:MFCD00013226
:256.94 g/mol
:356 °C
:220-882-2
:8,III
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Appearance:White to Light yellow powder to crystal
- Purity (Precipitation Titration):min. 98.0 %
Frequently Bought Together