
Trifluoromethanesulphonic acid ≥98.0% (by titrimetric analysis)
: TCI
:1493-13-6
:
:CHF₃O₃S
:MFCD00007514
:150.08 g/mol
:14,09676
:162°C
:35 °C
:1,69 g/cm³ (20 °C)
:216-087-5
:3265
:8,I
:GHS05
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Neutralization titration):min. 98.0 %
- Refractive index n20/D:1.3260 to 1.3310
Frequently Bought Together