Order Entry
Spain
ContactUsLinkComponent
Propyl hexanoate ≥98.0% (by GC)
 
Propyl hexanoate ≥98.0% (by GC)
  TCIAP1940-25ML
 :  TCI
 :  
undefined
 
Propyl hexanoate ≥98.0% (by GC)
  TCIAP1940-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :158.24 g/mol
 :187°C
 :-69°C
 :–69°C
 :0,867 g/cm³ (20 °C)
 :52°C
 :210-963-0
 :3272
 :3,III
 :GHS02
 :626-77-7
 :C₉H₁₈O₂
 :MFCD00053803