Sodium pyruvate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:113-24-6
:
Formula:CH₃COCOONa
MDL Number:MFCD00002586
:110.04 g/mol
Melting Pt:220…230 °C
Density:1,718 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:204-024-4
Specification Test Results
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together