Order Entry
Spain
ContactUsLinkComponent
Cobalt ≥99.8% (metals basis), nanopowder APS 25-30 nm
Flammable
Health
Cobalt ≥99.8% (metals basis), nanopowder APS 25-30 nm
Catalog # 46347.06
CAS Number:  
Flammable
Health
Cobalt ≥99.8% (metals basis), nanopowder APS 25-30 nm
Catalog # 46347.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :58.93 g/mol
Storage Temperature:Ambient
EINECS:231-158-0
UN:3089
ADR:4.1,II
 :GHS02
 :GHS08
CAS Number:7440-48-4
Formula:Co
MDL Number:MFCD00010935