Order Entry
Spain
ContactUsLinkComponent
trans-2,2-Dimethyl-3-heptene ≥99.0% (by GC)
Flammable
Health
trans-2,2-Dimethyl-3-heptene ≥99.0% (by GC)
Catalog # TCIAD1260-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Health
trans-2,2-Dimethyl-3-heptene ≥99.0% (by GC)
Catalog # TCIAD1260-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
 :126.24 g/mol
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–85 °C
Flash Pt:19°C
EINECS:243-156-7
UN:3295
ADR:3,II
 :GHS02
 :GHS08
CAS Number:19550-75-5
Formula:C₉H₁₈
MDL Number:MFCD00039961