
Triethyl phosphate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:78-40-0
:
Formula:C₆H₁₅O₄P
MDL Number:MFCD00009077
:182.16 g/mol
Merck Index:14,09674
Boiling Pt:216°C
Melting Pt:–57 °C
Density:1,07 g/cm³ (20 °C)
Flash Pt:111°C
EINECS:201-114-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together