Propyl formate ≥95.0% (by GC)
: TCI
: Formic acid propyl ester
:110-74-7
:
:C₄H₈O₂
:MFCD00003295
:81°C
:88.11 g/mol
:-93
:0°C
:14,07858
:203-798-0
:1281
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together