

Ethyl formate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:109-94-4
:
Formula:C₃H₆O₂
MDL Number:MFCD00003294
:74.08 g/mol
Merck Index:14,03807
Boiling Pt:54°C
Melting Pt:–80°C
Density:0,9208 g/cm³ (20 °C)
Flash Pt:–20°C
Storage Temperature:Ambient
EINECS:203-721-0
UN:1190
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together