D-(+)-Mannose ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
CAS Number:3458-28-4
:MFCD00799233
Formula:C₆H₁₂O₆
MDL Number:MFCD00064122
Boiling Pt:376
Molecular Weight:180.16 g/mol
Melting Pt:133
Merck Index:14,05747
Storage Temperature:Ambient
EINECS:222-392-4
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+14.0 to +15.5 deg (C=4, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together