Order Entry
Spain
ContactUsLinkComponent
 
 
p-Toluenesulphonyl chloride ≥99.0% (by GC)
 :  TCI
 
undefined
 
p-Toluenesulphonyl chloride ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 99.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    67.0 to 71.0 deg-C

 

Frequently Bought Together