


Benzocaine (ethyl-4-aminobenzoate) ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:94-09-7
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007892
:165.19 g/mol
Merck Index:14,01086
Boiling Pt:310°C
Melting Pt:89…90 °C
Flash Pt:112 °C
Storage Temperature:Ambient
EINECS:202-303-5
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:89.0 to 92.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together