Order Entry
Spain
ContactUsLinkComponent
1-Chloro-2-methylbutane ≥95.0% (by GC)
Flammable
1-Chloro-2-methylbutane ≥95.0% (by GC)
Catalog # TCIAC1459-5G
Supplier:  TCI
CAS Number:  
undefined
Flammable
1-Chloro-2-methylbutane ≥95.0% (by GC)
Catalog # TCIAC1459-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :106.6 g/mol
Boiling Pt:99°C
Flash Pt:0°C
EINECS:210-466-9
UN:1107
ADR:3,II
 :GHS02
CAS Number:616-13-7
Formula:C₅H₁₁Cl
MDL Number:MFCD00039366