Order Entry
Spain
ContactUsLinkComponent
Chloroacetaldehyde diethyl acetal ≥98.0% (by GC)
 
 
Chloroacetaldehyde diethyl acetal ≥98.0% (by GC)
  TCIAC0084-25G
 :  TCI
 :  
undefined
 
 
Chloroacetaldehyde diethyl acetal ≥98.0% (by GC)
  TCIAC0084-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :152.62 g/mol
 :157°C
 :–45 °C
 :1,0418 g/cm³ (0 °C)
 :29°C
 :210-695-4
 :1993
 :3,III
 :GHS02
 :GHS07
 :621-62-5
 :C₆H₁₃ClO₂
 :MFCD00000949