Order Entry
Spain
ContactUsLinkComponent
 
5-Bromo-2-chloroanisole ≥98.0% (by GC)
 :  TCI
 
undefined
5-Bromo-2-chloroanisole ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to lump
  • Purity (GC):
    min. 98.0 %
  • Freezing point:
    24.0 to 28.0 deg-C

 

Frequently Bought Together