(2-Chlorophenyl)acetic acid methyl ester ≥98.0% (by GC)
: TCI
:57486-68-7
:
:(C₆H₄Cl)CH₂COOCH₃
:MFCD00137445
:184.62 g/mol
:128°C
:260-764-8
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together