

N,N,N'-Trimethylethylenediamine ≥97.0% (by GC, titration analysis)
: TCI
:142-25-6
:
:C₅H₁₄N₂
:MFCD00014874
:102.18 g/mol
:116 °C (1013 hPa)
:0,783 g/cm³ (20 °C)
:14°C
:205-529-2
:2733
:3,II
:GHS02
:GHS05
- Appearance:Colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Frequently Bought Together