Order Entry
Spain
ContactUsLinkComponent
 
 
 
4-Fluoro-o-xylene ≥99.0% (by GC)
  TCIAF0355-10G
 :  TCI
 :  
undefined
 
 
4-Fluoro-o-xylene ≥99.0% (by GC)
  TCIAF0355-10G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :124.16 g/mol
 :60°C
 :1 g/cm³ (20 °C)
 :40°C
 :1993
 :3,III
 :GHS02
 :GHS07
 :452-64-2
 :C₈H₉F
 :MFCD00039217