Dibutyl sebacate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:109-43-3
:
Formula:C₁₈H₃₄O₄
MDL Number:MFCD00027218
:314.47 g/mol
Boiling Pt:345°C
Melting Pt:–10°C
Density:0,94 g/cm³ (20 °C)
Flash Pt:190°C
EINECS:203-672-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together