
Naphthacene ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:92-24-0
:
Formula:C₁₈H₁₂
MDL Number:MFCD00003702
:228.29 g/mol
Merck Index:14,06369
Boiling Pt:437 °C (1013 hPa)
Melting Pt:357°C
Density:1,323 g/cm³ (20 °C)
EINECS:202-138-9
:GHS08
Specification Test Results
- Appearance:Light yellow to Brown powder to crystaline
- Purity (HPLC):min. 97.0 area%
Specification Test Results
Related Information
Frequently Bought Together