
1,4-Diphenylbutadiyne ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:886-66-8
:
Formula:(C₆H₅)CCCC(C₆H₅)
MDL Number:MFCD00004787
:202.26 g/mol
Boiling Pt:210°C
Melting Pt:87 °C
Density:1,163 g/cm³ (20 °C)
EINECS:212-953-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:86.0 to 89.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together