
Allyl phenyl ether ≥95.0% (by GC)
: TCI
:1746-13-0
:
:H₂CCHCH₂OC₆H₅
:MFCD00008644
:134.18 g/mol
:195°C
:0,994 g/cm³ (0 °C)
:62°C
:217-125-3
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together