
2-Methylbenzylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89-93-0
:
Formula:C₈H₁₁N
MDL Number:MFCD00008112
:121.18 g/mol
Boiling Pt:82°C
Melting Pt:< 0 °C
Density:0,9921 g/cm³ (0 °C)
Flash Pt:84°C
EINECS:201-952-1
UN:2735
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together