Hydrazinium monochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Hydrazinium monochloride, Hydrazinium chloride, Hydrazine monohydrochloride, Hydrazine hydrochloride
CAS Number:2644-70-4
:
Formula:H₅ClN₂
MDL Number:MFCD00044368
Molecular Weight:68.51 g/mol
EINECS:220-154-4
UN:3290
ADR:6.1 II
:GHS05
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Potassium iodate Method):min. 98.0 %
- Melting point:92.0 to 95.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together