
Anthranilamide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:88-68-6
:
Formula:C₇H₈N₂O
MDL Number:MFCD00007981
:136.15 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:105…107 °C
Flash Pt:≥ 185 °C
Storage Temperature:Ambient
EINECS:201-851-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:110.0 to 113.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together