
3,5-Dichloro-4-hydroxybenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3336-41-2
:
Formula:C₇H₄Cl₂O₃
MDL Number:MFCD00002550
:207.01 g/mol
Boiling Pt:341 °C (1013 hPa)
Melting Pt:268…269 °C
EINECS:222-071-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together