Order Entry
Spain
ContactUsLinkComponent
2',7'-Dichlorofluorescein, Supelco®
2',7'-Dichlorofluorescein, Supelco®
Catalog # 1.09676.0005
Supplier:  Merck
CAS Number:  
2',7'-Dichlorofluorescein, Supelco®
Catalog # 1.09676.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    401.2 g/mol
  • Bulk Density:
    390 kg/m3

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :401.20 g/mol
EINECS:200-968-6
CAS Number:76-54-0
Formula:C₂₀H₁₀Cl₂O₅
MDL Number:MFCD00005047