Order Entry
Spain
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Benzo(k)fluoranthene solution
Catálogo # 49476-U
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Benzo(k)fluoranthene solution
Catálogo # 49476-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methylene chloride
  • Pk:
    1
  • Standards:
    Analytical standard
  • CAS No.:
    207-08-9
  • Description:
    Benzo(k)fluoranthene solution
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H319 - H336 - H351
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00046287
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Benzo[k]fluoranthene solution
  • Molecular weight:
    252,31

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P201 - P202 - P261 - P302 + P352 - P305 + P351 + P338 - P308 + P313