Cerotic acid
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:506-46-7
:
Formula:C₂₆H₅₂O₂
MDL Number:MFCD00002811
:396.7 g/mol
Boiling Pt:419 °C (1013 hPa)
Melting Pt:76…77 °C
Density:0,8198 g/cm³ (100 °C)
Storage Temperature:Refrigerator
EINECS:208-040-2
Frequently Bought Together