Order Entry
Spain
ContactUsLinkComponent
Synthetic leupeptin
Synthetic leupeptin
Certificates
Synthetic leupeptin

Specifications

  • CAS No.:
    103476-89-7
  • Protein/peptide type:
    Synthetic
  • Protein synonyms:
    Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate
  • Protein/peptide name:
    Leupeptin
  • Purity:
    90%
  • Molecular formula:
    C₄₀H₇₈N₁₂O₁₂S

Specifications

Related Information

Frequently Bought Together