Order Entry
Spain
ContactUsLinkComponent
Auramine O, Sigma-Aldrich®
Toxic
Auramine O, Sigma-Aldrich®
Catalog # A9655-25G
CAS Number:  
undefined
Toxic
Auramine O, Sigma-Aldrich®
Catalog # A9655-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :303.84 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:219-567-2
UN:2811
 :GHS06
Storage Temperature:Ambient
CAS Number:2465-27-2
Formula:C₁₇H₂₂ClN₃
MDL Number:MFCD00012484