Order Entry
Spain
ContactUsLinkComponent
Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide), Calbiochem®
Irritant
Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide), Calbiochem®
Catalog # 256455-1
CAS Number:  
Irritant
Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide), Calbiochem®
Catalog # 256455-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :862.05 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:86303-23-3
Formula:C₄₂H₇₅N₃O₁₅
MDL Number:MFCD00161482