

5-Bromo-2,3'-bipyridyl 95%
Catalog # H27880.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:235.08 g/mol
Melting Pt:75…78 °C
:GHS06
:GHS07
Storage Temperature:Ambient
CAS Number:774-53-8
Formula:C₁₀H₇BrN₂
MDL Number:MFCD09037420