Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fast Green FCF
Fast green FCF, Certifiable Grade
Fast green FCF, Certifiable Grade
Fast green FCF, Certifiable Grade
E772-25G
:
VWR Chemicals
:
Fast green FCF, Certifiable Grade
E772-25G
:
VWR Chemicals
:
Pack type:
Bottle, amber
Pk:
25 g
Seq:
3308
:
:
808.86 g/mol
:
13,03970
:
290 °C
:
219-091-5
:
GHS08
:
Ambient
:
2353-45-9
:
C₃₇H₃₄N₂Na₂O₁₀S₃
:
MFCD00013053