Order Entry
Spain
ContactUsLinkComponent
 
Pesticide reference standard, Amitraz 10 µg/ml in acetonitrile
Catalog # PROONE7041A
CAS Number:  
Pesticide reference standard, Amitraz 10 µg/ml in acetonitrile
Catalog # PROONE7041A
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    33089-61-1
  • Description:
    Amitraz 10 µg/ml in acetonitrile
  • Formula:
    C₁₉H₂₃N₃
  • Description:
    Amitra

Specifications

About this item