Dipropyleneglycol (mixture of isomers) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:25265-71-8
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00004538
:134.18 g/mol
Flash Pt:138°C
EINECS:246-770-3
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomer)
Specification Test Results
Frequently Bought Together