Dipropyleneglycol (mixture of isomers) for synthesis, Sigma-Aldrich®
Catalog # 8.03265.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-32 °C
- Molar Mass:134.17 g/mol
- Boiling Point:231 - 235 °C (1013 hPa)
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:121 °C
- Refractive Index:1.441 (20 °C, 589 nm)
- Explosion Limit:2.9 - 12.6 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:6 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:310 °C DIN 51794
Specification Test Results
About this item
:
:134.18 g/mol
:M-Clarity™ quality level = MQ200
EINECS:246-770-3
CAS Number:25265-71-8
Formula:C₆H₁₄O₃
MDL Number:MFCD00051023