Dipropyleneglycol (mixture of isomers) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25265-71-8
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00051023
:134.18 g/mol
Boiling Pt:231…235 °C
Melting Pt:–32 °C
Density:1.022
Storage Temperature:Ambient
EINECS:246-770-3
Related Information
Frequently Bought Together