Order Entry
Spain
ContactUsLinkComponent
Dipropyleneglycol (mixture of isomers) 99%
Dipropyleneglycol (mixture of isomers) 99%
Certificates
undefined
Dipropyleneglycol (mixture of isomers) 99%
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4420 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together