Pesticide reference standard, Acephate 10 µg/ml in acetonitrile
Catalog # PROONE7001A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:30560-19-1
- Description:Acephate 10 µg/ml in acetonitrile
- Formula:C₄H₁₀NO₃PS
- Description:Acephate