Ethylmagnesium bromide 3 M Stabilised with diethyl ether
About this item
CAS No.: 925-90-6 Molecular Formula: C2H5BrMg Formula Weight: 133.28 Physical Form: Liquid Density: 1.020 Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow to brown Odour: Ether-like Melting Point: -116.3° Boiling Point: 34.6° UN No.: 3399 MDL No.: MFCD00000043
- CAS Number:925-90-6
- Formula:C₂H₅BrMg
- MDL Number:MFCD00000043
- Molecular Weight:133.27 g/mol
- Boiling Pt:34.6 °C
- Melting Pt:–116.3 °C
- Density:1.020
- Storage Temperature:Ambient
- EINECS:213-127-3
- UN:3399
- ADR:4.3,I
- :Packaged under argon in resealable ChemSeal™ bottles.
2 Options Available Below
Product Details & Documents
CAS No.: 925-90-6 Molecular Formula: C2H5BrMg Formula Weight: 133.28 Physical Form: Liquid Density: 1.020 Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow to brown Odour: Ether-like Melting Point: -116.3° Boiling Point: 34.6° UN No.: 3399 MDL No.: MFCD00000043
- CAS Number:925-90-6
- Formula:C₂H₅BrMg
- MDL Number:MFCD00000043
- Molecular Weight:133.27 g/mol
- Boiling Pt:34.6 °C
- Melting Pt:–116.3 °C
- Density:1.020
- Storage Temperature:Ambient
- EINECS:213-127-3
- UN:3399
- ADR:4.3,I
- :Packaged under argon in resealable ChemSeal™ bottles.
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



